Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt11
Entrez Gene ID
22411
Gene Name
wingless-type MMTV integration site family, member 11
Gene Aliases
-
Chromosome Location
Chr.7: 98834921 - 98854747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285792.1 NP_001272721.1 2 - 3 400 72
GenBank mRNA AK077828.1 - 1 - 2 97 72

View Full Product Details