The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199035.2 | NP_950200.2 | 12 - 13 | 1370 | 131 |
GenBank mRNA | AK141341.1 | - | 12 - 13 | 1367 | 131 |
AK166755.1 | - | 12 - 13 | 1365 | 131 | |
AK166897.1 | - | 12 - 13 | 1371 | 131 | |
BC061244.1 | - | 12 - 13 | 1359 | 131 |