Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg8
Entrez Gene ID
381903
Gene Name
asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase)
Gene Aliases
AI447372, Gm1089
Chromosome Location
Chr.7: 97371594 - 97392159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199035.2 NP_950200.2 3 - 4 396 78
GenBank mRNA AK141341.1 - 3 - 4 390 78
AK166755.1 - 3 - 4 391 78
AK166897.1 - 3 - 4 397 78
BC061244.1 - 3 - 4 385 78

View Full Product Details