Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg8
Entrez Gene ID
381903
Gene Name
asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase)
Gene Aliases
AI447372, Gm1089
Chromosome Location
Chr.7: 97371594 - 97392159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199035.2 NP_950200.2 7 - 8 795 84
GenBank mRNA AK141341.1 - 7 - 8 792 84
AK166755.1 - 7 - 8 790 84
AK166897.1 - 7 - 8 796 84
BC061244.1 - 7 - 8 784 84

View Full Product Details