The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199035.2 | NP_950200.2 | 9 - 10 | 1063 | 91 |
GenBank mRNA | AK141341.1 | - | 9 - 10 | 1060 | 91 |
AK166755.1 | - | 9 - 10 | 1058 | 91 | |
AK166897.1 | - | 9 - 10 | 1064 | 91 | |
BC061244.1 | - | 9 - 10 | 1052 | 91 |