The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291358.1 | NP_001278287.1 | 3 - 4 | 545 | 69 |
NM_145583.3 | NP_663558.1 | 3 - 4 | 411 | 69 | |
GenBank mRNA | AK134287.1 | - | 3 - 4 | 428 | 69 |
AK146442.1 | - | 3 - 4 | 369 | 69 | |
AK159933.1 | - | 3 - 4 | 386 | 69 | |
AK159950.1 | - | 3 - 4 | 384 | 69 | |
AK162960.1 | - | 4 - 5 | 359 | 69 | |
BC004794.1 | - | 3 - 4 | 365 | 69 |