Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dnajb13
- Entrez Gene ID
-
69387
- Gene Name
- DnaJ (Hsp40) related, subfamily B, member 13
- Gene Aliases
-
1700014P03Rik, Tsarg
- Chromosome Location
-
Chr.7: 100503011 - 100514815 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005984.1 |
- |
1 - 2 |
138 |
159 |
View Full Product Details