The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163182.1 | NP_001156654.1 | 4 - 5 | 541 | 74 |
NM_001163183.1 | NP_001156655.1 | 3 - 4 | 394 | 74 | |
NM_001163184.1 | NP_001156656.1 | 3 - 4 | 394 | 74 | |
NM_001163185.1 | NP_001156657.1 | 3 - 4 | 384 | 74 | |
NM_001163186.1 | NP_001156658.1 | 2 - 3 | 385 | 74 | |
NM_001163187.1 | NP_001156659.1 | 2 - 3 | 385 | 74 | |
NM_001291336.1 | NP_001278265.1 | 3 - 4 | 394 | 74 | |
NM_013746.3 | NP_038774.1 | 4 - 5 | 541 | 74 | |
GenBank mRNA | AF000272.1 | - | 4 - 5 | 512 | 74 |
AF071000.1 | - | 2 - 3 | 373 | 74 | |
AF100613.1 | - | 2 - 3 | 373 | 74 | |
AF101053.1 | - | 4 - 5 | 512 | 74 | |
AK005102.1 | - | 3 - 4 | 349 | 74 | |
AK141634.1 | - | - | 352 | 74 | |
AK158172.1 | - | 3 - 4 | 385 | 74 | |
AK164683.1 | - | 3 - 4 | 349 | 74 | |
BC024756.1 | - | 4 - 5 | 502 | 74 | |
BY255927.1 | - | 2 - 3 | 387 | 74 | |
CX201870.1 | - | 3 - 4 | 346 | 74 |