The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291354.1 | NP_001278283.1 | 3 - 4 | 1286 | 74 |
NM_007491.2 | NP_031517.2 | 3 - 4 | 1338 | 74 | |
NR_111935.1 | 4 - 5 | 1005 | 74 | ||
GenBank mRNA | AJ297548.1 | - | 3 - 4 | 495 | 74 |
AK132854.1 | - | 3 - 4 | 1339 | 74 | |
BC116823.1 | - | 3 - 4 | 1150 | 74 | |
BC116849.1 | - | 3 - 4 | 1150 | 74 | |
BC144825.1 | - | 3 - 4 | 1150 | 74 | |
BC144826.1 | - | 3 - 4 | 1098 | 74 | |
BC144836.1 | - | 4 - 5 | 817 | 74 | |
U60881.1 | - | - | 1171 | 74 |