Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insc
Entrez Gene ID
233752
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
3830422K02Rik, C730020D23
Chromosome Location
Chr.7: 114743677 - 114850380 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173767.3 NP_776128.2 7 - 8 1105 62
GenBank mRNA AK014452.1 - 6 - 7 866 62
AK050143.1 - 4 - 5 600 62
AK211253.1 - 1 - 2 53 62
BC026405.1 - 7 - 8 992 62
BC089586.1 - 4 - 5 609 62
BC139196.1 - 4 - 5 493 62
BC139197.1 - 4 - 5 493 62
DQ205645.1 - 6 - 7 818 62

View Full Product Details