Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sox6
Entrez Gene ID
20679
Gene Name
SRY (sex determining region Y)-box 6
Gene Aliases
AI987981, SOX-LZ
Chromosome Location
Chr.7: 115470872 - 116040484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025559.3 NP_001020730.1 10 - 11 1270 131
NM_001025560.2 NP_001020731.1 10 - 11 1384 131
NM_001277326.1 NP_001264255.1 10 - 11 1365 131
NM_001277327.1 NP_001264256.1 9 - 10 1425 131
NM_001277328.1 NP_001264257.1 8 - 9 1130 131
NM_011445.4 NP_035575.1 11 - 12 1418 131
GenBank mRNA AK029696.1 - 8 - 9 1058 131
AK030713.1 - 10 - 11 1385 131
AK048143.1 - 10 - 11 1271 131
AK084290.1 - 10 - 11 1426 131
AK132409.1 - 10 - 11 1331 131
BC067407.1 - 9 - 10 1422 131
D61689.1 - 8 - 9 1126 131
U32614.1 - 11 - 12 1422 131

View Full Product Details