The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276301.1 | NP_001263230.1 | 12 - 13 | 2015 | 82 |
NM_009667.3 | NP_033797.2 | 12 - 13 | 2134 | 82 | |
GenBank mRNA | AK133465.1 | - | 12 - 13 | 2001 | 82 |
BC007183.1 | - | 7 - 8 | 981 | 82 | |
BC040366.1 | - | 12 - 13 | 2134 | 82 | |
BC056380.1 | - | 12 - 13 | 1976 | 82 | |
D85596.1 | - | 12 - 13 | 2013 | 82 |