The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030566.2 | NP_085043.2 | 1 - 2 | 390 | 83 |
GenBank mRNA | AK089938.1 | - | 1 - 2 | 94 | 83 |
AK144836.1 | - | 1 - 2 | 76 | 83 | |
AK157832.1 | - | 1 - 2 | 51 | 83 | |
BC015287.1 | - | 1 - 2 | 398 | 83 | |
BU514356.1 | - | 1 - 2 | 390 | 83 |