The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145589.2 | NP_663564.2 | 1 - 2 | 362 | 136 |
GenBank mRNA | AK133884.1 | - | 1 - 2 | 327 | 136 |
AK139559.1 | - | 2 - 3 | 137 | 136 | |
AK145816.1 | - | 2 - 3 | 142 | 136 | |
AK151203.1 | - | 2 - 3 | 194 | 136 | |
BC055033.1 | - | 1 - 2 | 362 | 136 |