The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178029.3 | NP_821172.2 | 8 - 9 | 2692 | 58 |
GenBank mRNA | BC049883.1 | - | 1 - 2 | 80 | 58 |