Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March7
Entrez Gene ID
57438
Gene Name
membrane-associated ring finger (C3HC4) 7
Gene Aliases
Axo, Axot, Gtrgeo17
Chromosome Location
Chr.2: 60209892 - 60249211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020575.2 NP_065600.1 2 - 3 314 120
GenBank mRNA AF155739.1 - 2 - 3 314 120
AK034109.1 - - 339 120
AK143177.1 - 2 - 3 334 120
AK166666.1 - 2 - 3 321 120
AK167246.1 - 2 - 3 296 120
AK168555.1 - 2 - 3 317 120
BC025029.1 - 2 - 3 278 120

View Full Product Details