Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stx4a
Entrez Gene ID
20909
Gene Name
syntaxin 4A (placental)
Gene Aliases
Stx4, Syn-4, Syn4
Chromosome Location
Chr.7: 127841769 - 127849686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009294.3 NP_033320.1 6 - 7 721 83
GenBank mRNA AK145909.1 - 6 - 7 740 83
AK161971.1 - 6 - 7 714 83
BC005791.1 - 6 - 7 561 83
BC011491.1 - 6 - 7 721 83
BC052023.1 - 6 - 7 657 83
U76832.1 - 6 - 7 490 83

View Full Product Details