Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx4a
Entrez Gene ID
20909
Gene Name
syntaxin 4A (placental)
Gene Aliases
Stx4, Syn-4, Syn4
Chromosome Location
Chr.7: 127841769 - 127849686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009294.3 NP_033320.1 8 - 9 929 86
GenBank mRNA AK145909.1 - 8 - 9 948 86
AK161971.1 - 8 - 9 922 86
BC005791.1 - 8 - 9 769 86
BC011491.1 - 8 - 9 929 86
BC052023.1 - 8 - 9 865 86
U76832.1 - 8 - 9 698 86

View Full Product Details