The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021282.2 | NP_067257.1 | 8 - 9 | 1313 | 128 |
GenBank mRNA | BC013451.1 | - | 8 - 9 | 1313 | 128 |
BC042693.1 | - | 8 - 9 | 1292 | 128 | |
DQ838976.1 | - | 5 - 6 | 820 | 128 | |
DQ838977.1 | - | 3 - 4 | 292 | 128 | |
L11650.1 | - | 8 - 9 | 1406 | 128 | |
X62595.1 | - | 8 - 9 | 1298 | 128 |