Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc12a1
Entrez Gene ID
20495
Gene Name
solute carrier family 12, member 1
Gene Aliases
AI788571, D630042G03Rik, Nkcc2, mBSC1, urehr3
Chromosome Location
Chr.2: 125152600 - 125230002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079690.1 NP_001073158.1 23 - 24 3090 124
NM_183354.2 NP_899197.2 23 - 24 3090 124
GenBank mRNA AI875508.1 - 2 - 3 124
BC016888.1 - 22 - 23 3068 124
BC051100.1 - 16 - 17 1854 124
U94518.1 - 22 - 23 3037 124

View Full Product Details