Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc12a1
Entrez Gene ID
20495
Gene Name
solute carrier family 12, member 1
Gene Aliases
AI788571, D630042G03Rik, Nkcc2, mBSC1, urehr3
Chromosome Location
Chr.2: 125152600 - 125230002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079690.1 NP_001073158.1 25 - 26 3315 122
NM_183354.2 NP_899197.2 25 - 26 3315 122
GenBank mRNA BC016888.1 - 24 - 25 3293 122
BC051100.1 - 18 - 19 2079 122
U20973.1 - 25 - 26 3313 122
U20974.1 - 25 - 26 3313 122
U20975.1 - 25 - 26 3313 122

View Full Product Details