Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dut
Entrez Gene ID
110074
Gene Name
deoxyuridine triphosphatase
Gene Aliases
5031412I06Rik, 5133400F09Rik, D2Bwg0749e, Dutp, dUTPase
Chromosome Location
Chr.2: 125247248 - 125259049 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159646.1 NP_001153118.1 1 - 2 160 86
GenBank mRNA AK017216.1 - 1 - 2 150 86
AK135438.1 - 1 - 2 169 86
BC019979.1 - 1 - 2 142 86

View Full Product Details