The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172100.1 | NP_001165571.1 | 8 - 9 | 1243 | 70 |
NM_001172101.1 | NP_001165572.1 | 8 - 9 | 1246 | 70 | |
NM_145135.3 | NP_660117.2 | 8 - 9 | 1394 | 70 | |
GenBank mRNA | AF071546.1 | - | 7 - 8 | 1114 | 70 |
AK133822.1 | - | 8 - 9 | 1174 | 70 | |
AK141351.1 | - | 8 - 9 | 1219 | 70 | |
AK145173.1 | - | 8 - 9 | 1228 | 70 | |
AK147059.1 | - | 8 - 9 | 1180 | 70 | |
BC010331.1 | - | 8 - 9 | 1263 | 70 |