Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rbbp6
- Entrez Gene ID
-
19647
- Gene Name
- retinoblastoma binding protein 6
- Gene Aliases
-
4933422O15Rik, AI316869, BB233631, C030034J04Rik, C77662, P2P-R, PACT
- Chromosome Location
-
Chr.7: 122970546 - 123002572 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011247.2 |
NP_035377.2 |
2 - 3 |
667 |
88 |
View Full Product Details