The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040137.2 | NP_001035227.1 | 2 - 3 | 264 | 86 |
GenBank mRNA | AK015115.1 | - | 2 - 3 | 265 | 86 |
AK015578.1 | - | 2 - 3 | 265 | 86 |