Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Paox
- Entrez Gene ID
-
212503
- Gene Name
- polyamine oxidase (exo-N4-amino)
- Gene Aliases
-
2410012F02Rik, AI118225, Pao
- Chromosome Location
-
Chr.7: 140115534 - 140134368 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC033913.1 |
- |
1 - 2 |
44 |
77 |
View Full Product Details