The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010183.1 | NP_034313.1 | 4 - 5 | 234 | 54 |
GenBank mRNA | BC027193.1 | - | 2 - 3 | 105 | 54 |
CF170568.1 | - | 3 - 4 | 196 | 54 |