Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbrs
Entrez Gene ID
14123
Gene Name
fibrosin
Gene Aliases
Fbs, Fbs1
Chromosome Location
Chr.7: 127477246 - 127491513 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010183.1 NP_034313.1 8 - 9 529 75
GenBank mRNA AK037952.1 - 4 - 5 308 75
BC027193.1 - 6 - 7 400 75
CF170568.1 - 7 - 8 491 75
CF532387.1 - 2 - 3 199 75

View Full Product Details