Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtg1
Entrez Gene ID
212508
Gene Name
mitochondrial GTPase 1 homolog (S. cerevisiae)
Gene Aliases
Gm169, Gtpbp7
Chromosome Location
Chr.7: 140137564 - 140150786 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199301.2 NP_955005.2 1 - 2 205 81
GenBank mRNA AK140018.1 - 1 - 2 121 81
AK144453.1 - 1 - 2 130 81
AK144625.1 - 1 - 2 163 81
BC027306.1 - 1 - 2 114 81
BY240360.1 - 1 - 2 205 81

View Full Product Details