Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syce1
Entrez Gene ID
74075
Gene Name
synaptonemal complex central element protein 1
Gene Aliases
4933406J07Rik
Chromosome Location
Chr.7: 140777229 - 140787854 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143765.1 NP_001137237.1 9 - 10 639 72
GenBank mRNA AK016694.1 - 9 - 10 639 72

View Full Product Details