The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029821.2 | NP_084097.1 | 1 - 2 | 263 | 87 |
GenBank mRNA | AK004470.1 | - | 1 - 2 | 481 | 87 |
AK013117.1 | - | 1 - 2 | 265 | 87 | |
BC051545.1 | - | 1 - 2 | 431 | 87 | |
DQ225768.1 | - | 1 - 2 | 138 | 87 |