The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033319.2 | NP_001028491.1 | 18 - 19 | 2556 | 65 |
NM_001195086.1 | NP_001182015.1 | 19 - 20 | 2631 | 65 | |
GenBank mRNA | BC038349.1 | - | 7 - 8 | 919 | 65 |
BC157936.1 | - | 19 - 20 | 2448 | 65 | |
BC158007.1 | - | 18 - 19 | 2513 | 65 | |
BC172096.1 | - | 19 - 20 | 2451 | 65 | |
EU568869.1 | - | 19 - 20 | 2631 | 65 |