The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289722.1 | NP_001276651.1 | 3 - 4 | 379 | 156 |
NM_001289723.1 | NP_001276652.1 | 3 - 4 | 344 | 156 | |
NM_153138.4 | NP_694778.1 | 3 - 4 | 374 | 156 | |
GenBank mRNA | AJ437262.1 | - | 2 - 3 | 175 | 156 |
AK031046.1 | - | 3 - 4 | 341 | 156 | |
AK156547.1 | - | 3 - 4 | 379 | 156 | |
BC049788.1 | - | 3 - 4 | 297 | 156 |