Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr.2: 144250123 - 144270902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199188.1 NP_001186117.1 4 - 5 496 89
NM_024225.5 NP_077187.1 3 - 4 407 89
GenBank mRNA AK007676.1 - 3 - 4 355 89
AK029463.1 - 3 - 4 367 89
AK031873.1 - 4 - 5 388 89
AK049129.1 - 4 - 5 388 89
AK078776.1 - 4 - 5 389 89
AK144710.1 - 4 - 5 389 89
AK152311.1 - 4 - 5 432 89
AK159904.1 - 3 - 4 312 89
AK169545.1 - 3 - 4 628 89
AK169786.1 - 4 - 5 388 89
AK184438.1 - 2 - 3 220 89
AK187575.1 - 2 - 3 219 89
BC002242.1 - 4 - 5 465 89

View Full Product Details