Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntsr2
Entrez Gene ID
18217
Gene Name
neurotensin receptor 2
Gene Aliases
NT2R, NTRL
Chromosome Location
Chr.12: 16653452 - 16660236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK005368.1 - 2 - 3 589 87

View Full Product Details