Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfml2a
Entrez Gene ID
241327
Gene Name
olfactomedin-like 2A
Gene Aliases
4932431K08Rik, mFLJ00237
Chromosome Location
Chr.2: 38931980 - 38960585 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172854.2 NP_766442.1 3 - 4 577 78
GenBank mRNA AK030069.1 - - 398 78
AK035313.1 - 3 - 4 578 78
AK220202.1 - 2 - 3 388 78
BC137871.1 - 3 - 4 505 78

View Full Product Details