The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290784.1 | NP_001277713.1 | 5 - 6 | 933 | 72 |
NM_178886.3 | NP_849217.2 | 5 - 6 | 936 | 72 | |
GenBank mRNA | AK029683.1 | - | 5 - 6 | 834 | 72 |
AK032208.1 | - | 5 - 6 | 822 | 72 | |
BC138177.1 | - | 5 - 6 | 936 | 72 | |
BC138178.1 | - | 6 - 7 | 934 | 72 | |
BC145174.1 | - | 5 - 6 | 932 | 72 |