Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galk2
Entrez Gene ID
69976
Gene Name
galactokinase 2
Gene Aliases
2810017M24Rik, AI327030, Gk2
Chromosome Location
Chr.2: 125859109 - 125984298 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291002.1 NP_001277931.1 2 - 3 251 72
NM_175154.3 NP_780363.1 2 - 3 357 72
GenBank mRNA AK053002.1 - 2 - 3 207 72
AK082521.1 - 2 - 3 248 72
AK154762.1 - 2 - 3 206 72
AK171939.1 - 2 - 3 234 72
AK176923.1 - 2 - 3 134 72
AK209145.1 - 2 - 3 72
BC079843.1 - 2 - 3 221 72

View Full Product Details