Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a27
Entrez Gene ID
74011
Gene Name
solute carrier family 25, member 27
Gene Aliases
3632410G24Rik, 9430092A03Rik, D530043E16Rik, Ucp4
Chromosome Location
Chr.17: 43638210 - 43667194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028711.3 NP_082987.1 7 - 8 804 86
GenBank mRNA AB106930.1 - 7 - 8 948 86
AK014394.1 - 7 - 8 805 86
AK043831.1 - 7 - 8 1004 86
AK219158.1 - 2 - 3 127 86
BC138993.1 - 7 - 8 811 86
BC138994.1 - 7 - 8 811 86

View Full Product Details