Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhb
Entrez Gene ID
211482
Gene Name
EF hand domain family, member B
Gene Aliases
4921525D22Rik
Chromosome Location
Chr.17: 53398048 - 53463334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172497.3 NP_766085.2 2 - 3 955 85
GenBank mRNA AK029550.1 - 2 - 3 956 85
BC132103.1 - 2 - 3 946 85
BC137880.1 - 2 - 3 946 85

View Full Product Details