Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhb
Entrez Gene ID
211482
Gene Name
EF hand domain family, member B
Gene Aliases
4921525D22Rik
Chromosome Location
Chr.17: 53398048 - 53463334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172497.3 NP_766085.2 9 - 10 1831 68
GenBank mRNA AK029550.1 - 9 - 10 1832 68
AK167235.1 - 6 - 7 1459 68
BC132103.1 - 9 - 10 1822 68
BC137880.1 - 9 - 10 1822 68

View Full Product Details