Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Treml2
- Entrez Gene ID
-
328833
- Gene Name
- triggering receptor expressed on myeloid cells-like 2
- Gene Aliases
-
AW049306, Gm750, Tlt2
- Chromosome Location
-
Chr.17: 48299387 - 48312534 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013294.1 |
- |
2 - 3 |
566 |
93 |
View Full Product Details