The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144939.2 | NP_659188.1 | 2 - 3 | 207 | 117 |
GenBank mRNA | BC014819.1 | - | 2 - 3 | 207 | 117 |