Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frs3
Entrez Gene ID
107971
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
4930417B13Rik, AI449674, Frs2beta, Snt2
Chromosome Location
Chr.17: 47687296 - 47704286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144939.2 NP_659188.1 3 - 4 297 64
GenBank mRNA BC014819.1 - 3 - 4 297 64

View Full Product Details