Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cobll1
Entrez Gene ID
319876
Gene Name
Cobl-like 1
Gene Aliases
1810047P18Rik, Coblr1, D430044D16Rik
Chromosome Location
Chr.2: 65088339 - 65239480 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027225.1 NP_081501.1 9 - 10 1678 158
GenBank mRNA AK037010.1 - 11 - 12 1783 158
AK052533.1 - 9 - 10 1627 158
AK173076.1 - 9 - 10 1509 158
BC067007.1 - 9 - 10 1675 158

View Full Product Details