The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027225.1 | NP_081501.1 | 1 - 2 | 264 | 112 |
NM_177025.5 | NP_795999.2 | 1 - 2 | 264 | 112 | |
GenBank mRNA | AK052533.1 | - | 1 - 2 | 213 | 112 |
AK052939.1 | - | 2 - 3 | 195 | 112 | |
AK164624.1 | - | 2 - 3 | 194 | 112 | |
AY308746.1 | - | 1 - 2 | 264 | 112 |