Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cerkl
Entrez Gene ID
228094
Gene Name
ceramide kinase-like
Gene Aliases
Gm1958, Rp26
Chromosome Location
Chr.2: 79331105 - 79430520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048176.1 NP_001041641.1 2 - 3 532 150
GenBank mRNA BC046474.1 - 2 - 3 638 150
BY742285.1 - 2 - 3 542 150
HQ426680.1 - 2 - 3 532 150
HQ426681.1 - 2 - 3 532 150
HQ426685.1 - 2 - 3 532 150
HQ426686.1 - 2 - 3 532 150
HQ426687.1 - 2 - 3 663 150
HQ426688.1 - 2 - 3 532 150
HQ426691.1 - - 331 150
HQ426692.1 - - 462 150
HQ426693.1 - - 372 150
HQ426694.1 - - 331 150
HQ426695.1 - - 504 150
HQ426696.1 - - 504 150
HQ426697.1 - - 504 150
HQ426698.1 - - 504 150
HQ426699.1 - - 504 150
HQ426700.1 - - 635 150
HQ426701.1 - - 504 150

View Full Product Details