Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndst2
Entrez Gene ID
17423
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2
Gene Aliases
Mndns, NDST-2
Chromosome Location
Chr.14: 20723730 - 20734621 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA U02304.1 - 1 - 2 213 90

View Full Product Details