The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007474.2 | NP_031500.1 | 1 - 2 | 171 | 92 |
GenBank mRNA | AF018952.1 | - | 1 - 2 | 153 | 92 |