Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zdhhc5
Entrez Gene ID
228136
Gene Name
zinc finger, DHHC domain containing 5
Gene Aliases
1110032A17Rik, AI451382, Zisp
Chromosome Location
Chr.2: 84687920 - 84715164 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144887.4 NP_659136.1 11 - 12 3212 63
GenBank mRNA AK028737.1 - 8 - 9 1778 63
AK075641.1 - 1 - 2 558 63
AK082513.1 - 11 - 12 3137 63
AK145086.1 - 11 - 12 3140 63
AK173251.1 - 11 - 12 3167 63
AY894891.1 - 11 - 12 3204 63
BC020051.1 - 10 - 11 2182 63
BC027047.1 - 7 - 8 1446 63
BC065155.1 - 11 - 12 3082 63

View Full Product Details